Gene Transcripts

Gene Symbol
Kcnk4
Entrez Gene ID
16528
Gene Name
potassium channel, subfamily K, member 4
Gene Aliases
MLZ-622, TRAAK, TRAAKt, Tex40
Chromosome Location
Chr. 19: 6925662 - 6935372 on Build GRCm38
UniGene ID
Mm.12894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008431.2 NP_032457.1 4 707
GenBank mRNA AF056492.1 - 4 745
BC119784.1 - 4 577 40106379

View Full Product Details