Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271603.1 | NP_001258532.1 | 2 | 854 | |
NM_009896.2 | NP_034026.1 | 2 | 734 | ||
GenBank mRNA | AB000677.1 | - | 1 | 578 | |
AB000710.1 | - | 1 | 632 | ||
AF120490.1 | - | 2 | 707 | ||
AF180302.1 | - | 2 | 712 | ||
AK028632.1 | - | 735 | |||
AK154706.1 | - | 751 | |||
BC132366.1 | - | 2 | 731 | 40130643 | |
BC132368.1 | - | 2 | 731 | 40130645 | |
U88325.1 | - | 2 | 696 |