Gene Transcripts

Gene Symbol
Dhh
Entrez Gene ID
13363
Gene Name
desert hedgehog
Gene Aliases
C78960
Chromosome Location
Chr. 15: 98881630 - 98898572 on Build GRCm38
UniGene ID
Mm.324688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007857.4 NP_031883.1 2 635
GenBank mRNA AK032466.1 - 2 618
AK033392.1 - 2 636
AK134149.1 - 2 1293

View Full Product Details