Gene Transcripts

Gene Symbol
Arl11
Entrez Gene ID
219144
Gene Name
ADP-ribosylation factor-like 11
Gene Aliases
ARLTS1, C730007L20Rik
Chromosome Location
Chr. 14: 61306288 - 61311937 on Build GRCm38
UniGene ID
Mm.206566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177337.3 NP_796311.2 2 705
GenBank mRNA AK008804.3 - 2 614
AK050053.1 - 2 616
AK155765.1 - 1 1327
BC064093.1 - 2 705 864589

View Full Product Details