Gene Transcripts

Gene Symbol
Lrrc41
Entrez Gene ID
230654
Gene Name
leucine rich repeat containing 41
Gene Aliases
AA409966, AW555107, D630045E04Rik, D730026A16Rik, MUF1
Chromosome Location
Chr. 4: 116075465 - 116097043 on Build GRCm38
UniGene ID
Mm.260786
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK052762.1 - 4 408
AK085717.1 - 4 499
AK154613.1 - 4 554
AK163171.1 - 4 501
AK170641.1 - 4 540
BC005765.1 - 3 276
BC022976.1 - 4 486 5362925
BC024940.1 - 4 484
BC071258.1 - 4 419

View Full Product Details