Gene Transcripts

Gene Symbol
2210011C24Rik
Entrez Gene ID
70134
Gene Name
RIKEN cDNA 2210011C24 gene
Gene Aliases
Chromosome Location
Chr. 8: 84010228 - 84011720 on Build GRCm38
UniGene ID
Mm.41017
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291292.1 NP_001278221.1 1 132
GenBank mRNA AI853297.1 - 1 180

View Full Product Details