Gene Transcripts

Gene Symbol
Sipa1l1
Entrez Gene ID
246212
Gene Name
signal-induced proliferation-associated 1 like 1
Gene Aliases
SPAL, Spa-1, Spa1, Spar
Chromosome Location
Chr. 6: 106052212 - 106163249 on Build Rnor_6.0
UniGene ID
Rn.10835
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139330.1 NP_647546.1 2 1398
GenBank mRNA AF026504.1 - 2 1398
AY043226.1 - 1 961

View Full Product Details