Gene Transcripts

Gene Symbol
Olr1684
Entrez Gene ID
294151
Gene Name
olfactory receptor 1684
Gene Aliases
ratchr20-350709-351650_ORF
Chromosome Location
Chr. 20: 572454 - 573395 on Build Rnor_6.0
UniGene ID
Rn.142691
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000270.1 NP_001000270.1 1 469

View Full Product Details