Gene Transcripts

Gene Symbol
Trip11
Entrez Gene ID
314393
Gene Name
thyroid hormone receptor interactor 11
Gene Aliases
Chromosome Location
Chr. 6: 125741520 - 125812928 on Build Rnor_6.0
UniGene ID
Rn.92005
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191772.1 NP_001178701.1 12 3441
XM_006240469.2 XP_006240531.1 11 4041
XM_006240470.2 XP_006240532.1 11 4038

View Full Product Details