Gene Transcripts

Gene Symbol
Olr1250
Entrez Gene ID
405092
Gene Name
olfactory receptor 1250
Gene Aliases
Chromosome Location
Chr. 4: 1658278 - 1659210 on Build Rnor_6.0
UniGene ID
Rn.143520
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000806.1 NP_001000806.1 1 79

View Full Product Details