Gene Transcripts

Gene Symbol
Msln
Entrez Gene ID
60333
Gene Name
mesothelin
Gene Aliases
Chromosome Location
Chr. 10: 15119700 - 15129129 on Build Rnor_6.0
UniGene ID
Rn.18607
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031658.1 NP_113846.1 12 1373
GenBank mRNA D87351.1 - 12 1373

View Full Product Details