Gene Transcripts

Gene Symbol
Plrg1
Entrez Gene ID
60376
Gene Name
pleiotropic regulator 1
Gene Aliases
Chromosome Location
Chr. 2: 182040338 - 182056669 on Build Rnor_6.0
UniGene ID
Rn.44181
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021757.2 NP_068525.1 7, 8 677
GenBank mRNA AF128241.1 - 6, 7 584
BC087742.1 - 7, 8 677 7314208

View Full Product Details