Gene Transcripts

Gene Symbol
Chn2
Entrez Gene ID
84031
Gene Name
chimerin 2
Gene Aliases
Chromosome Location
Chr. 4: 84423708 - 84452989 on Build Rnor_6.0
UniGene ID
Rn.10521
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032084.1 NP_114473.1 7 711
GenBank mRNA L07494.1 - 7 711
X69489.1 - 7 711

View Full Product Details