Gene Transcripts

Gene Symbol
Magea11
Entrez Gene ID
302845
Gene Name
melanoma antigen family A, 11
Gene Aliases
Chromosome Location
Chr. X: 141847092 - 141855448 on Build Rnor_6.0
UniGene ID
Rn.123462
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013962.1 NP_001013984.1 3 522
GenBank mRNA BC083676.1 - 3 522

View Full Product Details