Gene Transcripts

Gene Symbol
Stub1
Entrez Gene ID
287155
Gene Name
STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase
Gene Aliases
Chip
Chromosome Location
Chr. 10: 15197754 - 15200035 on Build Rnor_6.0
UniGene ID
Rn.6504
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025625.2 NP_001020796.2 2 285
GenBank mRNA BC099240.1 - 2 232
CB568781.1 - 2 286

View Full Product Details