Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039691.2 | NP_001034780.1 | 2 | 158 | |
NM_057105.3 | NP_476446.2 | 1 | 120 | ||
GenBank mRNA | AF359279.1 | - | 1 | 120 | |
AF461737.1 | - | 1 | 128 | ||
AY435133.1 | - | 1 | 35 | ||
BC107931.1 | - | 1 | 131 | 7126208 | |
D83796.1 | - | 1 | 35 | ||
J02612.1 | - | 2 | 159 | ||
J05132.1 | - | 2 | 297 | ||
S70355.1 | - | 3 | 98 |