Gene Transcripts

Gene Symbol
Ppp1r14a
Entrez Gene ID
114004
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 14A
Gene Aliases
Cpi17
Chromosome Location
Chr. 1: 87228581 - 87232842 on Build Rnor_6.0
UniGene ID
Rn.73852
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130403.1 NP_569087.1 1 147
GenBank mRNA AF352572.1 - 1 147
AY050673.1 - 1 147
FQ230520.1 - 1 139

View Full Product Details