Gene Transcripts

Gene Symbol
Grp
Entrez Gene ID
171101
Gene Name
gastrin releasing peptide
Gene Aliases
Chromosome Location
Chr. 18: 61563458 - 61576818 on Build Rnor_6.0
UniGene ID
Rn.10930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133570.5 NP_598254.2 2 437
GenBank mRNA AW524119.1 - 2 443
BF567195.1 - 2 418
M31176.1 - 2 841

View Full Product Details