Gene Transcripts

Gene Symbol
Olr1743
Entrez Gene ID
294197
Gene Name
olfactory receptor 1743
Gene Aliases
MOR121-1, Or8, ratchr20-1445925-1444984_ORF
Chromosome Location
Chr. 20: 1845210 - 1846151 on Build Rnor_6.0
UniGene ID
Rn.142661
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001425.1 NP_001001425.1 1 108

View Full Product Details