Gene Transcripts

Gene Symbol
Lsm8
Entrez Gene ID
296913
Gene Name
LSM8 homolog, U6 small nuclear RNA associated
Gene Aliases
Chromosome Location
Chr. 4: 45555077 - 45560943 on Build Rnor_6.0
UniGene ID
Rn.14059
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106585.1 NP_001100055.1 2 186
GenBank mRNA BC167094.1 - 2 57
FQ211862.1 - 152
FQ212479.1 - 152

View Full Product Details