Gene Transcripts

Gene Symbol
RGD1305420
Entrez Gene ID
298072
Gene Name
similar to Nef associated protein 1
Gene Aliases
Nap1
Chromosome Location
Chr. 5: 61990820 - 62001065 on Build Rnor_6.0
UniGene ID
Rn.51866
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025652.1 NP_001020823.1 4 1015
XM_008763696.1 XP_008761918.1 4 1082
GenBank mRNA BC097981.1 - 4 1015

View Full Product Details