Gene Transcripts

Gene Symbol
Efhc1
Entrez Gene ID
301295
Gene Name
EF-hand domain (C-terminal) containing 1
Gene Aliases
Chromosome Location
Chr. 9: 27068505 - 27107713 on Build Rnor_6.0
UniGene ID
Rn.198826
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122947.2 NP_001116419.2 10 1888

View Full Product Details