Gene Transcripts

Gene Symbol
Rspo3
Entrez Gene ID
498997
Gene Name
R-spondin 3
Gene Aliases
RGD1563246
Chromosome Location
Chr. 1: 30682049 - 30765160 on Build Rnor_6.0
UniGene ID
Rn.15751
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100990.1 NP_001094460.1 1 109
GenBank mRNA EU030376.1 - 1 109

View Full Product Details