Gene Transcripts

Gene Symbol
Zfp259
Entrez Gene ID
500989
Gene Name
zinc finger protein 259
Gene Aliases
RGD1562173
Chromosome Location
Chr. 8: 50563047 - 50572621 on Build Rnor_6.0
UniGene ID
Rn.102011
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001137646.1 NP_001131118.1 13, 14 1282

View Full Product Details