Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
65157
Gene Name
angiomotin like 2
Gene Aliases
Lccp
Chromosome Location
Chr. 8: 111210252 - 111226217 on Build Rnor_6.0
UniGene ID
Rn.25101
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031717.1 NP_113905.1 2 336
XM_006243680.2 XP_006243742.1 2 438

View Full Product Details