Gene Transcripts

Gene Symbol
Pax8
Entrez Gene ID
81819
Gene Name
paired box 8
Gene Aliases
Chromosome Location
Chr. 3: 1527316 - 1586023 on Build Rnor_6.0
UniGene ID
Rn.10392
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031141.2 NP_112403.2 7 888
XM_006233646.2 XP_006233708.1 7 1988
XM_008761610.1 XP_008759832.1 6 2324
XM_008761611.1 XP_008759833.1 6 2325
XM_008761612.1 XP_008759834.1 4 2052
XM_008761613.1 XP_008759835.1 6 2320
GenBank mRNA BC081779.1 - 7 888
X94246.1 - 6 771

View Full Product Details