Gene Transcripts

Gene Symbol
IGF1
Entrez Gene ID
3479
Gene Name
insulin like growth factor 1
Gene Aliases
IGF-I, IGFI, MGF
Chromosome Location
Chr.12: 102395867 - 102481839 on Build GRCh38
Uni Gene
Hs.160562
Interrogated Sequence Translated Protein
RefSeq NM_000618.4 NP_000609.1
NM_001111283.2 NP_001104753.1
NM_001111284.1 NP_001104754.1
XM_017019259.1 XP_016874748.1
XM_017019260.1 XP_016874749.1
XM_017019261.1 XP_016874750.1
XM_017019262.1 XP_016874751.1
XM_017019263.1 XP_016874752.1
GenBank mRNA AB209184.1 -
AK312231.1 -
AL603526.1 -
AL710591.1 -
BC148266.1 -
BC152321.1 -
CR541861.1 -
ENST00000337514.0 -
M11568.1 -
M27544.1 -
M29644.1 -
M37484.1 -
U40870.1 -
X00173.1 -
X56773.1 -
X56774.1 -
X57025.1 -

View Full Product Details