Gene Transcripts

Gene Symbol
FGF16
Entrez Gene ID
8823
Gene Name
fibroblast growth factor 16
Gene Aliases
FGF-16, MF4
Chromosome Location
Chr.X: 77447675 - 77456522 on Build GRCh38
Uni Gene
Hs.666364
Interrogated Sequence Translated Protein
RefSeq NM_003868.2 NP_003859.1
GenBank mRNA AB009391.1 -

View Full Product Details