Gene Transcripts

Gene Symbol
VEGFC
Entrez Gene ID
7424
Gene Name
vascular endothelial growth factor C
Gene Aliases
Flt4-L, LMPH1D, VRP
Chromosome Location
Chr.4: 176683534 - 176792745 on Build GRCh38
Uni Gene
Hs.435215
Interrogated Sequence Translated Protein
RefSeq NM_005429.4 NP_005420.1
GenBank mRNA AK313879.1 -
AW294939.1 -
BC035212.1 -
BC063685.1 -
BF528469.1 -
CR541897.1 -
DA044965.1 -
U43142.1 -
U58111.1 -
X94216.1 -

View Full Product Details