Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000586.3 | NP_000577.2 |
XM_017008177.1 | XP_016863666.1 | |
GenBank mRNA | A14844.1 | - |
AB451325.1 | - | |
AB451454.1 | - | |
AF228636.1 | - | |
AF532913.1 | - | |
AY283686.1 | - | |
AY523040.1 | - | |
BC066254.1 | - | |
BC066255.1 | - | |
BC066256.1 | - | |
BC066257.1 | - | |
BC070338.1 | - | |
DQ231169.1 | - | |
DQ861285.1 | - | |
ENST00000226730.0 | - | |
S77834.1 | - | |
S77835.1 | - | |
S82692.1 | - | |
U25676.1 | - | |
V00564.1 | - | |
X01586.1 | - |