Gene Transcripts

Gene Symbol
CCL5
Entrez Gene ID
6352
Gene Name
C-C motif chemokine ligand 5
Gene Aliases
D17S136E, RANTES, SCYA5, SIS-delta, SISd, TCP228, eoCP
Chromosome Location
Chr.17: 35871491 - 35880373 on Build GRCh38
Uni Gene
Hs.514821
Interrogated Sequence Translated Protein
RefSeq NM_001278736.1 NP_001265665.1
NM_002985.2 NP_002976.2
GenBank mRNA AA443491.1 -
AA486131.1 -
AF043341.1 -
AF266753.1 -
AK312212.1 -
AW769950.1 -
BC008600.1 -
BG272739.1 -
CB111656.1 -
CD695497.1 -
CR980938.1 -
DQ230537.1 -
GD136908.1 -
HY265548.1 -
M21121.1 -

View Full Product Details