Gene Transcripts

Gene Symbol
FGF9
Entrez Gene ID
2254
Gene Name
fibroblast growth factor 9
Gene Aliases
FGF-9, GAF, HBFG-9, HBGF-9, SYNS3
Chromosome Location
Chr.13: 21671076 - 21704501 on Build GRCh38
Uni Gene
Hs.111
Interrogated Sequence Translated Protein
RefSeq NM_002010.2 NP_002001.1
XM_011534996.2 XP_011533298.1
GenBank mRNA AK095571.1 -
AK290792.1 -
BC069692.1 -
BC103978.1 -
BC103979.1 -
CR746503.1 -
D14838.1 -
ENST00000382353.0 -

View Full Product Details