Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
3592
Gene Name
interleukin 12A
Gene Aliases
CLMF, IL-12A, NFSK, NKSF1, P35
Chromosome Location
Chr.3: 159988836 - 159996019 on Build GRCh38
Uni Gene
Hs.673
Interrogated Sequence Translated Protein
RefSeq NM_000882.3 NP_000873.2
GenBank mRNA AF101062.1 -
AF180562.1 -
AW472810.1 -
BC035571.1 -
BC104982.1 -
BC104984.1 -
BG702253.1 -
ENST00000305579.0 -
M65271.1 -
M65291.1 -

View Full Product Details