Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000515.4 | NP_000506.2 |
NM_022559.3 | NP_072053.1 | |
NM_022560.3 | NP_072054.1 | |
GenBank mRNA | A00469.1 | - |
AF110644.1 | - | |
AF185611.1 | - | |
AV749353.1 | - | |
AV753220.1 | - | |
AY613431.1 | - | |
AY613432.1 | - | |
BC062475.1 | - | |
BC075012.2 | - | |
BC075013.2 | - | |
BC090045.1 | - | |
CD108592.1 | - | |
ENST00000323322.0 | - | |
M14398.1 | - | |
V00519.1 | - |