Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000207.2 | NP_000198.1 |
NM_001185097.1 | NP_001172026.1 | |
NM_001185098.1 | NP_001172027.1 | |
NM_001291897.1 | NP_001278826.1 | |
GenBank mRNA | AY899304.1 | - |
BC005255.1 | - | |
BM510347.1 | - | |
BM510748.1 | - | |
BP322143.1 | - | |
BT006808.1 | - | |
DQ778082.1 | - | |
ENST00000250971.0 | - | |
JF909299.1 | - | |
JQ951950.1 | - | |
X70508.1 | - |