Gene Transcripts

Gene Symbol
IL2
Entrez Gene ID
3558
Gene Name
interleukin 2
Gene Aliases
IL-2, TCGF, lymphokine
Chromosome Location
Chr.4: 122449479 - 122456495 on Build GRCh38
Uni Gene
Hs.89679
Interrogated Sequence Translated Protein
RefSeq NM_000586.3 NP_000577.2
XM_017008177.1 XP_016863666.1
GenBank mRNA A14844.1 -
AB451325.1 -
AB451454.1 -
AF228636.1 -
AF532913.1 -
AY283686.1 -
AY523040.1 -
BC066254.1 -
BC066255.1 -
BC066256.1 -
BC066257.1 -
BC070338.1 -
DQ231169.1 -
DQ861285.1 -
ENST00000226730.0 -
S77834.1 -
S77835.1 -
S82692.1 -
U25676.1 -
V00564.1 -
X01586.1 -

View Full Product Details