Gene Transcripts

Gene Symbol
CDKN1A
Entrez Gene ID
1026
Gene Name
cyclin dependent kinase inhibitor 1A
Gene Aliases
CAP20, CDKN1, CIP1, MDA-6, P21, SDI1, WAF1, p21CIP1
Chromosome Location
Chr.6: 36676460 - 36687339 on Build GRCh38
Uni Gene
Hs.370771
Interrogated Sequence Translated Protein
RefSeq NM_000389.4 NP_000380.1
NM_001220777.1 NP_001207706.1
NM_001220778.1 NP_001207707.1
NM_001291549.1 NP_001278478.1
NM_078467.2 NP_510867.1
GenBank mRNA AA534613.1 -
AB209881.1 -
AB451290.1 -
AB451422.1 -
AI973138.1 -
AK298901.1 -
AK309512.1 -
BC000275.1 -
BC000312.2 -
BC001935.1 -
BC013967.2 -
BQ941282.1 -
BT006719.1 -
BU154329.1 -
BU193165.1 -
CR536533.1 -
DA552678.1 -
DA582789.1 -
DA699672.1 -
DA702185.1 -
DA838987.1 -
ENST00000244741.0 -
L25610.1 -
L26165.1 -
L47232.1 -
L47233.1 -
S67388.1 -
U03106.1 -
U09579.1 -

View Full Product Details