Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000618.4 | NP_000609.1 |
NM_001111283.2 | NP_001104753.1 | |
NM_001111284.1 | NP_001104754.1 | |
XM_017019259.1 | XP_016874748.1 | |
XM_017019260.1 | XP_016874749.1 | |
XM_017019261.1 | XP_016874750.1 | |
XM_017019262.1 | XP_016874751.1 | |
XM_017019263.1 | XP_016874752.1 | |
GenBank mRNA | AB209184.1 | - |
AK312231.1 | - | |
AL603526.1 | - | |
AL710591.1 | - | |
BC148266.1 | - | |
BC152321.1 | - | |
CR541861.1 | - | |
ENST00000337514.0 | - | |
M11568.1 | - | |
M27544.1 | - | |
M29644.1 | - | |
M37484.1 | - | |
U40870.1 | - | |
X00173.1 | - | |
X56773.1 | - | |
X56774.1 | - | |
X57025.1 | - |