Gene Transcripts

Gene Symbol
HLA-DRB4
Entrez Gene ID
3126
Gene Name
major histocompatibility complex, class II, DR beta 4
Gene Aliases
DR-4, DR4, DRB4, HLA-DR4B
Chromosome Location
-
Uni Gene
Hs.534322
Interrogated Sequence Translated Protein
RefSeq NM_021983.4 NP_068818.4
GenBank mRNA AF017439.1 -
AF048707.1 -
AF361548.1 -
AF361549.1 -
AI829825.1 -
AK225683.1 -
AK225690.1 -
AK291499.1 -
AK292151.1 -
AK293020.1 -
AY961064.1 -
AY961067.1 -
AY961070.2 -
BC005312.1 -
BF974249.1 -
DQ284431.1 -
DQ284433.1 -
GQ891550.1 -
M14190.1 -
M15071.1 -
M15178.1 -
M15179.1 -
M16732.1 -
M16942.1 -
M17385.1 -
M17388.1 -
M30180.1 -
M59803.1 -
S48645.1 -
U17379.1 -
U29342.1 -
U35375.1 -
U35729.1 -
U37433.1 -
U47028.1 -
U70543.1 -
U70544.1 -

View Full Product Details