Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001322809.1 | NP_001309738.1 |
NM_015985.3 | NP_057069.1 | |
XM_011529239.2 | XP_011527541.1 | |
GenBank mRNA | AF074332.1 | - |
AF113708.1 | - | |
AK304756.1 | - | |
BC111976.1 | - | |
BC111978.1 | - | |
BM999095.1 | - | |
ENST00000381922.0 | - |