Gene Transcripts

Gene Symbol
INS
Entrez Gene ID
3630
Gene Name
insulin
Gene Aliases
IDDM, IDDM1, IDDM2, ILPR, IRDN, MODY10
Chromosome Location
Chr.11: 2159779 - 2161209 on Build GRCh38
Uni Gene
Hs.272259
Interrogated Sequence Translated Protein
RefSeq NM_000207.2 NP_000198.1
NM_001185097.1 NP_001172026.1
NM_001185098.1 NP_001172027.1
NM_001291897.1 NP_001278826.1
GenBank mRNA AY899304.1 -
BC005255.1 -
BM510347.1 -
BM510748.1 -
BP322143.1 -
BT006808.1 -
DQ778082.1 -
ENST00000250971.0 -
JF909299.1 -
JQ951950.1 -
X70508.1 -

View Full Product Details