Gene Transcripts

Gene Symbol
JAK3
Entrez Gene ID
3718
Gene Name
Janus kinase 3
Gene Aliases
JAK-3, JAK3_HUMAN, JAKL, L-JAK, LJAK
Chromosome Location
Chr.19: 17824782 - 17848071 on Build GRCh38
Uni Gene
Hs.515247
Interrogated Sequence Translated Protein
RefSeq NM_000215.3 NP_000206.2
XM_005259896.3 XP_005259953.1
XM_011527990.1 XP_011526292.1
XM_011527991.2 XP_011526293.1
GenBank mRNA AK296383.1 -
AK304392.1 -
BC028068.1 -
BF512748.1 -
BM561964.1 -
DA405504.1 -
ENST00000458235.0 -
KU178153.1 -
KU178154.1 -
U08340.1 -
U09607.1 -
U31317.1 -
U31601.1 -
U57096.1 -

View Full Product Details