Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_000948.5 | NP_000939.1 |
NM_001163558.2 | NP_001157030.1 | |
XM_011514753.2 | XP_011513055.1 | |
XM_011514754.2 | XP_011513056.1 | |
GenBank mRNA | BC015850.1 | - |
BC088370.1 | - | |
CD106243.1 | - | |
D00411.1 | - | |
DQ839492.1 | - | |
ENST00000306482.0 | - | |
GQ305133.1 | - | |
M29386.1 | - | |
M58594.1 | - | |
U75583.1 | - | |
V00566.1 | - | |
X54393.1 | - | |
X54394.1 | - |