Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_021983.4 | NP_068818.4 |
GenBank mRNA | AF017439.1 | - |
AF048707.1 | - | |
AF361548.1 | - | |
AF361549.1 | - | |
AI829825.1 | - | |
AK225683.1 | - | |
AK225690.1 | - | |
AK291499.1 | - | |
AK292151.1 | - | |
AK293020.1 | - | |
AY961064.1 | - | |
AY961067.1 | - | |
AY961070.2 | - | |
BC005312.1 | - | |
BF974249.1 | - | |
DQ284431.1 | - | |
DQ284433.1 | - | |
GQ891550.1 | - | |
M14190.1 | - | |
M15071.1 | - | |
M15178.1 | - | |
M15179.1 | - | |
M16732.1 | - | |
M16942.1 | - | |
M17385.1 | - | |
M17388.1 | - | |
M30180.1 | - | |
M59803.1 | - | |
S48645.1 | - | |
U17379.1 | - | |
U29342.1 | - | |
U35375.1 | - | |
U35729.1 | - | |
U37433.1 | - | |
U47028.1 | - | |
U70543.1 | - | |
U70544.1 | - |