Gene Transcripts

Gene Symbol
ANGPT4
Entrez Gene ID
51378
Gene Name
angiopoietin 4
Gene Aliases
ANG3, ANG4
Chromosome Location
Chr.20: 869899 - 916367 on Build GRCh38
Uni Gene
Hs.278973
Interrogated Sequence Translated Protein
RefSeq NM_001322809.1 NP_001309738.1
NM_015985.3 NP_057069.1
XM_011529239.2 XP_011527541.1
GenBank mRNA AF074332.1 -
AF113708.1 -
AK304756.1 -
BC111976.1 -
BC111978.1 -
BM999095.1 -
ENST00000381922.0 -

View Full Product Details