Gene Transcripts

Gene Symbol
ANKRD36BP2
Entrez Gene ID
645784
Gene Name
ankyrin repeat domain 36B pseudogene 2
Gene Aliases
-
Chromosome Location
Chr.2: 88765902 - 88806596 on Build GRCh38
Uni Gene
Hs.105323
Interrogated Sequence Translated Protein
RefSeq NR_015424.1
GenBank mRNA AK097649.1 -
AL137717.1 -
BC031698.1 -
BX537549.1 -
BX648045.1 -

View Full Product Details