Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_013653.3 | NP_038681.2 |
GenBank mRNA | AF065944.1 | - |
AF065945.1 | - | |
AF065946.1 | - | |
AF065947.1 | - | |
AF128187.1 | - | |
AK003101.1 | - | |
AK158074.1 | - | |
AY722103.1 | - | |
BC033508.1 | - | |
BE333205.1 | - | |
CT010315.1 | - | |
M77747.1 | - | |
S37648.1 | - |