Gene Transcripts

Gene Symbol
Il12a
Entrez Gene ID
16159
Gene Name
interleukin 12a
Gene Aliases
IL-12p35, Il-12a, Ll12a, p35
Chromosome Location
Chr.3: 68690644 - 68698550 on Build GRCm38
Uni Gene
Mm.103783
Interrogated Sequence Translated Protein
GenBank mRNA AF128210.1 -
AF128211.1 -
AF128212.1 -
AF128213.1 -
AK153729.1 -
AV363060.1 -
BC116851.1 -
BC116855.1 -
M86672.1 -

View Full Product Details