Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_013518.4 | NP_038546.2 |
GenBank mRNA | AI414273.1 | - |
AK041815.1 | - | |
AK079617.1 | - | |
AK158584.1 | - | |
AK158782.1 | - | |
AK164411.1 | - | |
BC099872.1 | - | |
BC099873.1 | - | |
BC099874.1 | - | |
BC099875.1 | - | |
BC125237.1 | - | |
BX512213.1 | - | |
D38258.1 | - | |
HM988990.1 | - | |
S82023.1 | - | |
U33535.1 | - |