Interrogated Sequence | Translated Protein | |
---|---|---|
RefSeq | NM_001111274.1 | NP_001104744.1 |
NM_001111275.1 | NP_001104745.1 | |
NM_001111276.1 | NP_001104746.1 | |
NM_010512.4 | NP_034642.2 | |
NM_184052.3 | NP_908941.1 | |
GenBank mRNA | AF440694.1 | - |
AK038119.1 | - | |
AK050118.1 | - | |
AK052033.1 | - | |
AK081019.1 | - | |
AK155435.1 | - | |
AK165471.1 | - | |
AY878192.1 | - | |
AY878193.1 | - | |
BC012409.1 | - | |
CA494190.1 | - | |
CT010364.1 | - | |
X04480.1 | - | |
X04482.1 | - |